missing translation for 'onlineSavingsMsg'
Learn More

2-Acetamido-4-(4-bromophenyl)-5-methylthiazole, 97%, Thermo Scientific™

Catalog Number 15474056
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
313364-84-0
C12H11BrN2OS
311.197
MFCD00518863
KHPHCLMGHXPKDD-UHFFFAOYSA-N
n-4-4-bromophenyl-5-methyl-1,3-thiazol-2-yl acetamide, 2-acetamido-4-4-bromophenyl-5-methylthiazole, n-4-4-bromophenyl-5-methylthiazol-2-yl acetamide
3505149
N-[4-(4-bromophenyl)-5-methyl-1,3-thiazol-2-yl]acetamide
CC1=C(N=C(S1)NC(=O)C)C2=CC=C(C=C2)Br
This item is not returnable. View return policy
313364-84-0
C12H11BrN2OS
311.197
MFCD00518863
KHPHCLMGHXPKDD-UHFFFAOYSA-N
n-4-4-bromophenyl-5-methyl-1,3-thiazol-2-yl acetamide, 2-acetamido-4-4-bromophenyl-5-methylthiazole, n-4-4-bromophenyl-5-methylthiazol-2-yl acetamide
3505149
N-[4-(4-bromophenyl)-5-methyl-1,3-thiazol-2-yl]acetamide
CC1=C(N=C(S1)NC(=O)C)C2=CC=C(C=C2)Br

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 5 g
Formula Weight 311.2
Percent Purity 97%
Chemical Name or Material 2-Acetamido-4-(4-bromophenyl)-5-methylthiazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'