missing translation for 'onlineSavingsMsg'
Läs mer

2-Acetamido-4,6-O-benzylidene-2-deoxy-D-glucopyranose, TRC

Produktkod. 30316938
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30316938 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30316938 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA1425001G

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material 2-Acetamido-4,6-O-benzylidene-2-deoxy-D-glucopyranose
CAS 29776-43-0
Molekylformel C15 H19 N O6
Kvantitet 1 g
InChI formel InChI=1S/C15H19NO6/c1-8(17)16-11-12(18)13-10(21-14(11)19)7-20-15(22-13)9-5-3-2-4-6-9/h2-6,10-15,18-19H,7H2,1H3,(H,16,17)/t10-,11-,12-,13-,14?,15?/m1/s1
LEDER CC(=O)N[C@H]1C(O)O[C@@H]2COC(O[C@H]2[C@@H]1O)c3ccccc3
IUPAC-namn N-[(4aR,7R,8R,8aS)-6,8-dihydroxy-2-phenyl-4,4a,6,7,8,8a-hexahydropyrano[3,2-d][1,3]dioxin-7-yl]acetamide
Molekylvikt (g/mol) 309.31
Formel vikt 309.1212
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.