Läs mer
2-Acetoxy-1,4-naphthoquinone, TRC
Produktkod. 30312296
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30312296 250 mg
1 options
Produktkod. 30312296 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA167658250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-Acetoxy-1,4-naphthoquinone
CAS 1785-65-5
Molekylformel C12H8O4
Kvantitet 250 mg
Synonym 2-(Acetyloxy)-1,4-naphthalenedione, 2-Hydroxy-1,4-naphthoquinone Acetate, 2-(Acetyloxy)-1,4-naphthalenedione, 2-Hydroxynaphthoquinone Acetate, Lawsone Acetate, NSC 100542
InChI formel InChI=1S/C12H8O4/c1-7(13)16-11-6-10(14)8-4-2-3-5-9(8)12(11)15/h2-6H,1H3
LEDER CC(=O)OC1=CC(=O)c2ccccc2C1=O
IUPAC-namn (1,4-dioxonaphthalen-2-yl) acetate
Molekylvikt (g/mol) 216.19
Formel vikt 216.0423