Läs mer
2-Acetyl-octanoic Acid Methyl Ester, TRC
Produktkod. 30318356
Change view
Kvantitet:
5 g
Produktkod. Kvantitet
30318356 5 g
1 options
Produktkod. 30318356 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA1875505G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-Acetyl-octanoic Acid Methyl Ester
CAS 70203-04-2
Molekylformel C11H20O3
Kvantitet 5 g
Synonym Methyl 2-Acetyloctanoate, Methyl 2-Hexylacetoacetate, Methyl 2-Hexylacetylacetonate, Methyl alpha-Hexylacetoacetate
InChI formel InChI=1S/C11H20O3/c1-4-5-6-7-8-10(9(2)12)11(13)14-3/h10H,4-8H2,1-3H3
LEDER CCCCCCC(C(=O)C)C(=O)OC
IUPAC-namn methyl 2-acetyloctanoate
Molekylvikt (g/mol) 200.28
Formel vikt 200.1412