Läs mer
2-(Acetylamino)-3-(phenylthio)propanoic Acid, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | 2-(Acetylamino)-3-(phenylthio)propanoic Acid |
| CAS | 20640-68-0 |
| Molekylformel | C11H13NO3S |
| Kvantitet | 500 mg |
| Synonym | Cysteine, N-acetyl-S-phenyl- (9CI, ACI), DL-Cysteine, N-acetyl-S-phenyl- (ZCI), Alanine, N-acetyl-3-(phenylthio)-, DL- (8CI), N-Acetyl-S-phenylcysteine (ACI), (±)-Phenylmercapturic acid, 2-Acetamido-3-(phenylsulfanyl)propanoic acid, 2-Acetamido-3-(phenylthio)propanoic acid, 2-Acetamido-3-phenylsulfanylpropanoic acid, DL-Phenylmercapturic acid, N-Acetyl-S-phenyl-DL-cysteine, NSC 337386 |
| InChI formel | InChI=1S/C11H13NO3S/c1-8(13)12-10(11(14)15)7-16-9-5-3-2-4-6-9/h2-6,10H,7H2,1H3,(H,12,13)(H,14,15) |
| LEDER | CC(NC(C(O)=O)CSC1=CC=CC=C1)=O |
| IUPAC-namn | 2-acetamido-3-phenylsulfanylpropanoic acid |
| Molekylvikt (g/mol) | 239.29 |
| Formel vikt | 239.0616 |