Läs mer
2-Acetylbutanoic-d5 Acid Ethyl Ester, TRC
Produktkod. 30325581
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30325581 250 mg
1 options
Produktkod. 30325581 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA171152250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-Acetyl-butanoic-d5 Acid Ethyl Ester
CAS 141327-44-8
Molekylformel C8 2H5 H9 O3
Kvantitet 250 mg
Synonym Butanoic-3,3,4,4,4-d5 acid, 2-acetyl-, ethyl ester, Ethyl 2-acetylbutanoate-3,3,4,4,4-d5, Ethyl 3-oxo-2-(pentadeuteroethyl)butanoate, 2-Acetylbutanoic-d5 acid ethyl ester, Ethyl 2-ethyl-3-oxobutanoate-d5, Ethyl 2-(ethyl-d5)-3-oxobutanoate, Ethyl 2-ethyl-3-oxobutanoate-D5 (2-ethyl-D5), 2-(Ethyl-d5)acetoacetic acid ethyl ester, Ethyl α-ethylacetoacetate-d5
InChI formel InChI=1S/C8H14O3/c1-4-7(6(3)9)8(10)11-5-2/h7H,4-5H2,1-3H3/i1D3,4D2
LEDER [2H]C([2H])([2H])C([2H])([2H])C(C(=O)C)C(=O)OCC
IUPAC-namn ethyl 2-acetyl-3,3,4,4,4-pentadeuteriobutanoate
Molekylvikt (g/mol) 163.23
Formel vikt 163.1257