Learn More
2-Acetylfluorene, TRC
Product Code. 30320794
Change view
Click to view available options
Kvantitet:
500 mg
Product Code. Kvantitet
30320794 500 mg
1 options
Product Code. 30320794 Supplier Toronto Research Chemicals Supplier No. TRCA175918500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Kemiskt namn eller material 2-Acetylfluorene
CAS 781-73-7
Molekylformel C15H12O
Kvantitet 500 mg
Synonym 1-(9H-Fluoren-2-yl)ethanone, 2-Acetofluorene, 2-Acetyl-9H-fluorene, 2-Acetylfluorene, 2-Fluorenyl Methyl Ketone, NSC 12351
InChI formel InChI=1S/C15H12O/c1-10(16)11-6-7-15-13(8-11)9-12-4-2-3-5-14(12)15/h2-8H,9H2,1H3
LEDER CC(=O)c1ccc2c(Cc3ccccc23)c1
IUPAC-namn 1-(9H-fluoren-2-yl)ethanone
Molekylvikt (g/mol) 208.25
Formel vikt 208.0888