Läs mer
2-(Acetyloxy)-1-phenylethanone, TRC
Produktkod. 30318520
Change view
Kvantitet:
5 g
Produktkod. Kvantitet
30318520 5 g
1 options
Produktkod. 30318520 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA1877705G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-(Acetyloxy)-1-phenylethanone
CAS 2243-35-8
Molekylformel C10H10O3
Kvantitet 5 g
Synonym 2-Acetoxyacetophenone, 2-Hydroxyacetophenoneacetate, Acetic Acid 2-oxo-2-phenylethyl Ester, Benzoylmethyl acetate, NSC 9837, Phenacyl acetate, alpha-Acetoxyacetophenone, ω-Acetoxyacetophenone
InChI formel InChI=1S/C10H10O3/c1-8(11)13-7-10(12)9-5-3-2-4-6-9/h2-6H,7H2,1H3
LEDER CC(=O)OCC(=O)c1ccccc1
IUPAC-namn phenacyl acetate
Molekylvikt (g/mol) 178.18
Formel vikt 178.063