Läs mer
2-(Acetyloxy)-3-nitrobenzoic Acid Methyl Ester, TRC
Produktkod. 30325939
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30325939 250 mg
1 options
Produktkod. 30325939 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA187855250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-(Acetyloxy)-3-nitrobenzoic Acid Methyl Ester
CAS 22621-42-7
Molekylformel C10 H9 N O6
Kvantitet 250 mg
Synonym 3-Nitrosalicylic Acid Methyl Ester Acetate
InChI formel InChI=1S/C10H9NO6/c1-6(12)17-9-7(10(13)16-2)4-3-5-8(9)11(14)15/h3-5H,1-2H3
LEDER COC(=O)c1cccc(c1OC(=O)C)[N+](=O)[O-]
IUPAC-namn methyl 2-acetyloxy-3-nitrobenzoate
Molekylvikt (g/mol) 239.182
Formel vikt 239.043