Learn More
Julabo™ 2-Adapters for Presto™ Temperature Control System
Brand:  Julabo™ 8890082
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 15204283
  • 3140.00 SEK / Pack of 2
Estimated Shipment: 04-06-2024
to see stock.
Specifications
Suitable for PRESTO A30, PRESTO A40, PRESTO A45, PRESTO A45t, PRESTO A80 Temperature Control System
2-Adapters
M30 x 1.5 Female to M38 x 1.5 Female