Learn More
Julabo™ 2-Adapters for Presto™ Temperature Control System
Brand:  Julabo™ 8890080
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 15284273
  • 2805.00 SEK / Pack of 2
Estimated Shipment: 12-06-2024
to see stock.
Specifications
Suitable for PRESTO W91, PRESTO W91t, PRESTO W91tt, PRESTO W91ttx, PRESTO W91tx Powerfell Temperature Control System
2-Adapters
M30 x 1.5 Female to M38 x 1.5 Male