missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-1-(4-nitrophenyl)ethan-1-one hydrochloride hydrate, 97%, Thermo Scientific™

Catalog Number p-2697663
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
5425-81-0
C8H9ClN2O3
216.621
MFCD00025591
UJBOVJRECBNSDI-UHFFFAOYSA-N
2-amino-1-4-nitrophenyl ethanone hydrochloride, 2-amino-4'-nitro acetophenone hydrochloride, 2-amino-p-nitro-acetophenone hcl, 2-amino-p-nitro-acetophenone hydrochloride, 2-amino-1-4-nitro-phenyl ethanone hydrochloride, 2-amino-1-4-nitro-phenyl-ethanone hydrochloride, ethanone, 2-amino-1-4-nitrophenyl-, monohydrochloride, 2-amino-1-4-nitrophenyl ethan-1-one, chloride, 4-nitrophenacylamine hydrochloride hydrate, aronis24405
2792801
2-amino-1-(4-nitrophenyl)ethanone;hydrochloride
C1=CC(=CC=C1C(=O)CN)[N+](=O)[O-].Cl
This item is not returnable. View return policy
5425-81-0
C8H9ClN2O3
216.621
MFCD00025591
UJBOVJRECBNSDI-UHFFFAOYSA-N
2-amino-1-4-nitrophenyl ethanone hydrochloride, 2-amino-4'-nitro acetophenone hydrochloride, 2-amino-p-nitro-acetophenone hcl, 2-amino-p-nitro-acetophenone hydrochloride, 2-amino-1-4-nitro-phenyl ethanone hydrochloride, 2-amino-1-4-nitro-phenyl-ethanone hydrochloride, ethanone, 2-amino-1-4-nitrophenyl-, monohydrochloride, 2-amino-1-4-nitrophenyl ethan-1-one, chloride, 4-nitrophenacylamine hydrochloride hydrate, aronis24405
2792801
2-amino-1-(4-nitrophenyl)ethanone;hydrochloride
C1=CC(=CC=C1C(=O)CN)[N+](=O)[O-].Cl
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula HCl·H2O
Quantity 250 mg
Formula Weight 234.64
Percent Purity 97%
Chemical Name or Material 2-Amino-1-(4-nitrophenyl)ethan-1-one hydrochloride hydrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'