Läs mer
2-Amino-1-hydroxyethane-1,1-diphosphonic Acid, TRC
Produktkod. 30312481
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30312481 25 mg
1 options
Produktkod. 30312481 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA62798025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-Amino-1-hydroxyethane-1,1-diphosphonic Acid
CAS 41003-10-5
Molekylformel C2 H9 N O7 P2
Kvantitet 25 mg
Synonym P,P'-(2-amino-1-hydroxyethylidene)bisphosphonic acid, Phosphonic acid, (2-amino-1-hydroxyethylidene)bis- (9CI), 2-Amino-1-hydroxyethane-1,1-diphosphonic acid, 2-Amino-1-hydroxyethylene-1,1-bisphosphonic acid, Ethane-1-hydroxy-2-amino-1,1-diphosphonic acid, Glycine Bisphosphonate
InChI formel InChI=1S/C2H9NO7P2/c3-1-2(4,11(5,6)7)12(8,9)10/h4H,1,3H2,(H2,5,6,7)(H2,8,9,10)
LEDER NCC(O)(P(=O)(O)O)P(=O)(O)O
IUPAC-namn (2-amino-1-hydroxy-1-phosphonoethyl)phosphonic acid
Molekylvikt (g/mol) 221.04
Formel vikt 220.9854