Läs mer
2-Amino-2-[2-(4-heptylphenyl)ethyl]-1,3-propanediol Hydrochloride (Fingolimod Heptyl Analogue), Mikromol™
Produktkod. 17933766
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
17933766 25 mg
1 options
Produktkod. 17933766 Leverantör Mikromol Leverantörsnummer MM3554.040025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2-Amino-2-[2-(4-heptylphenyl)ethyl]-1,3-propanediol Hydrochloride (Fingolimod Heptyl Analogue)
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-Amino-2-[2-(4-heptylphenyl)ethyl]-1,3-propanediol Hydrochloride (Fingolimod Heptyl Analogue)
CAS 162361-44-6
Analyt- eller komponentnamn 2-Amino-2-[2-(4-heptylphenyl)ethyl]-1,3-propanediol Hydrochloride (Fingolimod Heptyl Analogue)
Frakt skick Room Temperature
Molekylformel C18 H31 N O2 . Cl H
Certifikat och efterlevnad ISO 17025
Synonym 1,3-Propanediol, 2-amino-2-[2-(4-heptylphenyl)ethyl]-, hydrochloride (1:1), 1,3-Propanediol, 2-amino-2-[2-(4-heptylphenyl)ethyl]-, hydrochloride (9CI), 2-Amino-2-[2-(4-heptylphenyl)ethyl]-1,3-propanediol hydrochloride (1:1), Fingolimod Hydrochloride Impurity B as Hydrochloride, Fingolimod Impurity B as Hydrochloride
InChI formel InChI=1S/C18H31NO2.ClH/c1-2-3-4-5-6-7-16-8-10-17(11-9-16)12-13-18(19,14-20)15-21;/h8-11,20-21H,2-7,12-15,19H2,1H3;1H
LEDER Cl.CCCCCCCc1ccc(CCC(N)(CO)CO)cc1
IUPAC-namn 2-amino-2-[2-(4-heptylphenyl)ethyl]propane-1,3-diol;hydrochloride
Molekylvikt (g/mol) 329.91
Kvantitet 25 mg
Formel vikt 329.2122 g/mol