Läs mer
2-Amino-3,4-dimethyl-3H-imidazo[4,5-f]quinoline, TRC
Produktkod. 18072310
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18072310 25 mg
18000420 10 mg
18000730 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 18072310 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA60520025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 77094-11-2
Molekylformel C12 H12 N4
Molekylvikt (g/mol) 212.25
Synonym 3,4-Dimethyl-3H-imidazo[4,5-f]quinolin-2-amine, 2-Amino-3,4-dimethylimidazo[4,5-f]quinoline, MeIQ, Methyl-IQ
IUPAC-namn 3,4-dimethylimidazo[4,5-f]quinolin-2-amine
LEDER Cc1cc2ncccc2c3nc(N)n(C)c13
Specifikationer
Kemiskt namn eller material 2-Amino-3,4-dimethyl-3H-imidazo[4,5-f]quinoline
Anteckningar om renhetsgrad HPLC
Kvantitet 25 mg
InChI formel InChI=1S/C12H12N4/c1-7-6-9-8(4-3-5-14-9)10-11(7)16(2)12(13)15-10/h3-6H,1-2H3,(H2,13,15)
Formel vikt 212.1062
Procent renhet >95%