missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-fluorobenzonitrile, 95%, Thermo Scientific Chemicals

Catalog Number p-7053853
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
115661-37-5
C7H5FN2
136.129
UNISSOLHERSZOW-UHFFFAOYSA-N
2783392
2-amino-3-fluorobenzonitrile
C1=CC(=C(C(=C1)F)N)C#N
This item is not returnable. View return policy
115661-37-5
C7H5FN2
136.129
UNISSOLHERSZOW-UHFFFAOYSA-N
2783392
2-amino-3-fluorobenzonitrile
C1=CC(=C(C(=C1)F)N)C#N

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
UN Number UN3439
Synonym 2-cyano-6-fluoroaniline, benzonitrile,2-amino-3-fluoro, benzonitrile, 2-amino-3-fluoro, acmc-1c91u, ksc493s2f, 2-amino-3-fluorobenzenecarbonitrile
Formula Weight 136.13
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2-Amino-3-fluorobenzonitrile

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'