Läs mer
2-Amino-3-hydroxyphenazine, Mikromol™
Produktkod. 17977194
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17977194 25 mg
17958054 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17977194 Leverantör Mikromol Leverantörsnummer MM2500.020025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2-Amino-3-hydroxyphenazine
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-Amino-3-hydroxyphenazine
CAS 4569-77-1
Analyt- eller komponentnamn 2-Amino-3-hydroxyphenazine
Frakt skick Room Temperature
Molekylformel C12 H9 N3 O
Certifikat och efterlevnad ISO 9001
Synonym 2-Amino-3-hydroxyphenazine
InChI formel InChI=1S/C12H9N3O/c13-7-5-10-11(6-12(7)16)15-9-4-2-1-3-8(9)14-10/h1-6,16H,13H2
LEDER Nc1cc2nc3ccccc3nc2cc1O
IUPAC-namn 3-aminophenazin-2-ol
Molekylvikt (g/mol) 211.22
Kvantitet 25 mg
Formel vikt 211.0746 g/mol