Läs mer
2-Amino-3-methyl-3H-imidazo[4,5-f]quinoline-2-13C, TRC
Produktkod. 30135672
Change view
Kvantitet:
5 mg
50 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135672 500 mg
30133362 50 mg
30006415 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30135672 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA616600500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 210049-11-9
Molekylformel 13C C10 H10 N4
Molekylvikt (g/mol) 199.22
Synonym 3H-Imidazo[4,5-f]quinolin-2-amine-2-13C, 3-methyl- (9CI), 2-Amino-3-methyl-3H-imidazo(4,5-f)quinoline-2-13C
IUPAC-namn 3-methyl(2^{13}C)imidazolo[4,5-f]quinolin-2-amine
LEDER Cn1[13c](N)nc2c3cccnc3ccc12
Specifikationer
Kemiskt namn eller material 2-Amino-3-methyl-3H-imidazo[4,5-F]quinoline-2-13C
Kvantitet 500 mg
InChI formel InChI=1S/C11H10N4/c1-15-9-5-4-8-7(3-2-6-13-8)10(9)14-11(15)12/h2-6H,1H3,(H2,12,14)/i11+1
Formel vikt 199.0939