Läs mer
2-Amino-3-methyl-9H-pyrido[2,3-b]indole-d3, TRC
Produktkod. 30326216
Change view
Kvantitet:
0.5 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30326216 5 mg
30316973 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30326216 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA6175025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-Amino-3-methyl-9H-pyrido[2,3-b]indole-d3
CAS 1189458-14-7
Molekylformel C12 D3 H8 N3
Kvantitet 5 mg
Synonym 2-Amino-3-methyl-α-carboline-d3, 3-Methyl-9H-1,9-diazafluoren-2-amine-d3
InChI formel InChI=1S/C12H11N3/c1-7-6-9-8-4-2-3-5-10(8)14-12(9)15-11(7)13/h2-6H,1H3,(H3,13,14,15)/i1D3
LEDER [2H]C([2H])([2H])c1cc2c3ccccc3[nH]c2nc1N
IUPAC-namn 3-(trideuteriomethyl)-9H-pyrido[2,3-b]indol-2-amine
Molekylvikt (g/mol) 200.2543
Formel vikt 200.1141