Läs mer
2-Amino-3-methyl-9H-pyrido[2,3-b]indole, TRC
Produktkod. 18082530
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18082530 10 mg
18040940 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18082530 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA61750010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 68006-83-7
Molekylformel C12 H11 N3
Molekylvikt (g/mol) 197.24
Synonym 3-Methyl-1H-pyrido[2,3-b]indol-2-amine, 1H-Pyrido[2,3-b]indol-2-amine, 3-methyl- (9CI), 3-Methyl-9H-pyrido[2,3-b]indol-2-amine, 2-Amino-3-methyl-9H-pyrido[2,3-b]indole, 2-Amino-3-methyl-α-carboline, 3-Methyl-9H-1,9-diazafluoren-2-amine, MeAαC, MeAaC
IUPAC-namn 3-methyl-9H-pyrido[2,3-b]indol-2-amine
LEDER Cc1cc2c3ccccc3[nH]c2nc1N
Specifikationer
Kemiskt namn eller material 2-Amino-3-methyl-9H-pyrido[2,3-b]indole
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C12H11N3/c1-7-6-9-8-4-2-3-5-10(8)14-12(9)15-11(7)13/h2-6H,1H3,(H3,13,14,15)
Formel vikt 197.0953
Procent renhet >95%