Läs mer
2-Amino-3-methylbutanamide Hydrochloride, TRC
Produktkod. 30008662
Change view
Kvantitet:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30008662 100 mg
30005424 1 g
30006696 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30008662 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA614630100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 93169-29-0
Molekylformel C5 H12 N2 O . H Cl
Molekylvikt (g/mol) 152.622
Synonym DL-Valinamide Hydrochloride, Butanamide, 2-amino-3-methyl-, hydrochloride (1:1), Butanamide, 2-amino-3-methyl-, monohydrochloride (9CI), Butanamide, 2-amino-3-methyl-, monohydrochloride, (±)-, Valinamide, hydrochloride (7CI), DL-Valinamide hydrochloride, 2-Amino-3-methylbutanamide hydrochloride
IUPAC-namn 2-amino-3-methyl-butanamide;hydrochloride
LEDER Cl.CC(C)C(N)C(=O)N
Specifikationer
Kemiskt namn eller material 2-Amino-3-methylbutanamide Hydrochloride
InChI formel InChI=1S/C5H12N2O.ClH/c1-3(2)4(6)5(7)8;/h3-4H,6H2,1-2H3,(H2,7,8);1H
Kvantitet 100 mg
Formel vikt 152.0716