missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-methylpyridine, 97%, Thermo Scientific Chemicals

Catalog Number p-7080622
230.37 SEK valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1603-40-3
C6H8N2
108.144
MFCD00006320
RGDQRXPEZUNWHX-UHFFFAOYSA-N
2-amino-3-picoline, 2-amino-3-methylpyridine, 2-pyridinamine, 3-methyl, 3-methyl-2-pyridinamine, 3-methyl-2-aminopyridine, 3-methyl-2-pyridylamine, 3-picoline, 2-amino, 2-amino-3-methyl pyridine, 2-amino-beta-picoline, pyridine, 2-amino-3-methyl
15347
3-methylpyridin-2-amine
CC1=C(N=CC=C1)N
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1603-40-3
C6H8N2
108.144
MFCD00006320
RGDQRXPEZUNWHX-UHFFFAOYSA-N
2-amino-3-picoline, 2-amino-3-methylpyridine, 2-pyridinamine, 3-methyl, 3-methyl-2-pyridinamine, 3-methyl-2-aminopyridine, 3-methyl-2-pyridylamine, 3-picoline, 2-amino, 2-amino-3-methyl pyridine, 2-amino-beta-picoline, pyridine, 2-amino-3-methyl
15347
3-methylpyridin-2-amine
CC1=C(N=CC=C1)N

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 29°C to 32°C
Density 1.073
Boiling Point 221°C to 222°C
Flash Point 118°C (244°F)
Odor Unpleasant
Refractive Index 1.5825
Quantity 25 g
UN Number UN2811
Beilstein 107892
Sensitivity Hygroscopic
Formula Weight 108.14
Percent Purity 97%
Chemical Name or Material 2-Amino-3-methylpyridine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'