missing translation for 'onlineSavingsMsg'
Learn More

2-amino-3-pyridinecarboxaldehyde 97%, Thermo Scientific™

Catalog Number p-3211591
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy
7521-41-7
C6H6N2O
122.127
MFCD01830382
NXMFJCRMSDRXLD-UHFFFAOYSA-N
2-aminonicotinaldehyde, 2-amino-3-pyridinecarboxaldehyde, 2-amino-3-formylpyridine, 3-pyridinecarboxaldehyde, 2-amino, 2-amino-pyridine-3-carbaldehyde, 2-aminopyridine-3-carboxaldehyde, 2-amino-3-pyridine carboxaldehyde, 2-amino-pyridin-3-carbaldehyde, pubchem1262, 2-amino nicotinaldehyde
737633
2-aminopyridine-3-carbaldehyde
C1=CC(=C(N=C1)N)C=O
This item is not returnable. View return policy
7521-41-7
C6H6N2O
122.127
MFCD01830382
NXMFJCRMSDRXLD-UHFFFAOYSA-N
2-aminonicotinaldehyde, 2-amino-3-pyridinecarboxaldehyde, 2-amino-3-formylpyridine, 3-pyridinecarboxaldehyde, 2-amino, 2-amino-pyridine-3-carbaldehyde, 2-aminopyridine-3-carboxaldehyde, 2-amino-3-pyridine carboxaldehyde, 2-amino-pyridin-3-carbaldehyde, pubchem1262, 2-amino nicotinaldehyde
737633
2-aminopyridine-3-carbaldehyde
C1=CC(=C(N=C1)N)C=O

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 95.0°C to 102.0°C
Color Yellow to Brown
Flash Point >300°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in chloroform
Formula Weight 122.13
Percent Purity 97%
Physical Form Powder, Crystals or Chunks
Chemical Name or Material 2-Amino-3-pyridinecarboxaldehyde
Show More Show Less
compliance-icons
Product Identifier
  • 2-Amino-3-pyridinecarboxaldehyde
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'