missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-(2-naphthyl)thiazole, 98%, Thermo Scientific Chemicals

Catalog Number p-7026912
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
21331-43-1
C13H10N2S
226.297
MFCD00046452
GWDNDNTTXIIXRS-UHFFFAOYSA-N
4-naphthalen-2-yl-thiazol-2-ylamine, 4-naphthalen-2-yl-1,3-thiazol-2-amine, 4-2-naphthyl-1,3-thiazol-2-amine, 2-amino-4-2-naphthyl thiazole, 4-naphthalen-2-yl thiazol-2-amine, 4-naphthalen-2-yl-thiazol-2-yl-amine, 4-2-naphthalenyl-2-thiazolamine, 2-thiazolamine,4-2-naphthalenyl, 4-2-naphthyl-1,3-thiazole-2-ylamine, acmc-20ao57
673702
4-naphthalen-2-yl-1,3-thiazol-2-amine
C1=CC=C2C=C(C=CC2=C1)C3=CSC(=N3)N
This item is not returnable. View return policy
21331-43-1
C13H10N2S
226.297
MFCD00046452
GWDNDNTTXIIXRS-UHFFFAOYSA-N
4-naphthalen-2-yl-thiazol-2-ylamine, 4-naphthalen-2-yl-1,3-thiazol-2-amine, 4-2-naphthyl-1,3-thiazol-2-amine, 2-amino-4-2-naphthyl thiazole, 4-naphthalen-2-yl thiazol-2-amine, 4-naphthalen-2-yl-thiazol-2-yl-amine, 4-2-naphthalenyl-2-thiazolamine, 2-thiazolamine,4-2-naphthalenyl, 4-2-naphthyl-1,3-thiazole-2-ylamine, acmc-20ao57
673702
4-naphthalen-2-yl-1,3-thiazol-2-amine
C1=CC=C2C=C(C=CC2=C1)C3=CSC(=N3)N

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 153°C to 156°C
Quantity 1 g
Beilstein 173533
Formula Weight 226.3
Percent Purity 98%
Chemical Name or Material 2-Amino-4-(2-naphthyl)thiazole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'