Läs mer
2-Amino-4,5,7,8-tetrahydro-6-(N-carbethoxy)thiazolo[5,4-d]azepine, TRC
Produktkod. 30323566
Change view
Kvantitet:
25 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30323566 50 mg
30324116 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30323566 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA62992050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-Amino-4,5,7,8-tetrahydro-6-(N-carbethoxy)thiazolo[5,4-d]az
CAS 887352-57-0
Molekylformel C10 H15 N3 O2 S
Kvantitet 50 mg
InChI formel InChI=1S/C10H15N3O2S/c1-2-15-10(14)13-5-3-7-8(4-6-13)16-9(11)12-7/h2-6H2,1H3,(H2,11,12)
LEDER CCOC(=O)N1CCc2nc(N)sc2CC1
IUPAC-namn ethyl 2-amino-4,5,7,8-tetrahydro-[1,3]thiazolo[4,5-d]azepine-6-carboxylate
Molekylvikt (g/mol) 241.31
Formel vikt 241.0885