Läs mer
2-Amino-4,6-dihydroxy-5-nitropyrimidine, TRC
Produktkod. 30328682
Change view
Kvantitet:
500 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30328682 500 mg
30326253 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30328682 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA604910500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-Amino-4,6-dihydroxy-5-nitropyrimidine
CAS 80466-56-4
Molekylformel C4 H4 N4 O4
Kvantitet 500 mg
InChI formel InChI=1S/C4H4N4O4/c5-4-6-2(9)1(8(11)12)3(10)7-4/h(H4,5,6,7,9,10)
LEDER Nc1nc(O)c(c(O)n1)[N+](=O)[O-]
IUPAC-namn 2-amino-5-nitropyrimidine-4,6-diol
Molekylvikt (g/mol) 172.1
Formel vikt 172.0233