Läs mer
2-Amino-4'-hydroxyacetophenone hydrochloride, TRC
Produktkod. 30309999
Change view
Kvantitet:
5 g
Produktkod. Kvantitet
30309999 5 g
1 options
Produktkod. 30309999 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA6087755G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-Amino-4'-hydroxyacetophenone hydrochloride
CAS 19745-72-3
Molekylformel C8H9NO2 . HCl
Kvantitet 5 g
Synonym 2-Amino-1-(4-hydroxyphenyl)ethanone Hydrochloride, 2-Amino-4'-Hydroxyacetophenone Hydrochloride, ?-Amino-4-hydroxyacetophenone Hydrochloride, ?-Amino-p-hydroxyacetophenone Hydrochloride
InChI formel InChI=1S/C8H9NO2.ClH/c9-5-8(11)6-1-3-7(10)4-2-6;/h1-4,10H,5,9H2;1H
LEDER O=C(C1=CC=C(C=C1)O)CN.Cl
IUPAC-namn 2-amino-1-(4-hydroxyphenyl)ethan-1-one hydrochloride
Molekylvikt (g/mol) 187.63
Formel vikt 187.04