missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-hydroxypyridine, 98%, Thermo Scientific Chemicals

Catalog Number p-7056086
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
33631-05-9
C5H6N2O
110.116
MFCD04114161
HQNIMNQVKVPZES-UHFFFAOYSA-N
2-amino-4-hydroxypyridine, 2-aminopyridin-4-ol, 2-amino-4-pyridinol, 4-pyridinol, 2-amino, 2-amino-pyridin-4-ol, zlchem 31, pubchem6677, acmc-1acz5, 4-hydroxypyridine-2-amine, 2-aminopyridin-4 1h-one
820936
2-amino-1H-pyridin-4-one
C1=CNC(=CC1=O)N
This item is not returnable. View return policy
33631-05-9
C5H6N2O
110.116
MFCD04114161
HQNIMNQVKVPZES-UHFFFAOYSA-N
2-amino-4-hydroxypyridine, 2-aminopyridin-4-ol, 2-amino-4-pyridinol, 4-pyridinol, 2-amino, 2-amino-pyridin-4-ol, zlchem 31, pubchem6677, acmc-1acz5, 4-hydroxypyridine-2-amine, 2-aminopyridin-4 1h-one
820936
2-amino-1H-pyridin-4-one
C1=CNC(=CC1=O)N

Employed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Employed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field

Solubility
Slightly soluble in water.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Color Yellow
Quantity 1 g
Beilstein 385930
Solubility Information Slightly soluble in water.
Formula Weight 110.12
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Amino-4-hydroxypyridine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'