Läs mer
2-Amino-4-isopropylamino-6-chlorotriazine, TRC
Produktkod. 30308942
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30308942 250 mg
1 options
Produktkod. 30308942 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA611750250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Atrazine-desethyl
CAS 6190-65-4
Molekylformel C6 H10 Cl N5
Kvantitet 250 mg
Synonym 1,3,5-Triazine-2,4-diamine, 6-chloro-N-(1-methylethyl)- (9CI), s-Triazine, 2-amino-4-chloro-6-(isopropylamino)- (6CI,7CI,8CI), 6-Chloro-N2-(1-methylethyl)-1,3,5-triazine-2,4-diamine, 2-Amino-4-chloro-6-(isopropylamino)-s-triazine, 2-Chloro-4-amino-6-isopropylamino-1,3,5-triazine, 2-Chloro-4-amino-6-isopropylamino-s-triazine, 2-Chloro-4-isopropylamino-6-amino-s-triazine, 2-Isopropylamino-4-amino-6-chloro-1,3,5-triazine, 6-Amino-2-chloro-4-isopropylamino-s-triazine, Atrazine-desethyl, DEA, Deethylatratone, Deethylatrazine, Desethylatrazine, G 30033, N-Deethylatrazine, s-Chloroaminoisopropylaminotriazine
InChI formel InChI=1S/C6H10ClN5/c1-3(2)9-6-11-4(7)10-5(8)12-6/h3H,1-2H3,(H3,8,9,10,11,12)
LEDER CC(C)Nc1nc(N)nc(Cl)n1
IUPAC-namn 6-chloro-2-N-propan-2-yl-1,3,5-triazine-2,4-diamine
Molekylvikt (g/mol) 187.63
Formel vikt 187.0625