Läs mer
2-Amino-4-methoxy-3-nitropyridine, TRC
Produktkod. 30308918
Change view
Kvantitet:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30308918 100 mg
30315591 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30308918 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA611947100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-Amino-4-methoxy-3-nitropyridine
CAS 84487-08-1
Molekylformel C6 H7 N3 O3
Kvantitet 100 mg
Synonym 4-Methoxy-3-nitro-2-pyridinamine, 2-Amino-4-methoxy-3-nitropyridine
InChI formel InChI=1S/C6H7N3O3/c1-12-4-2-3-8-6(7)5(4)9(10)11/h2-3H,1H3,(H2,7,8)
LEDER COc1ccnc(N)c1[N+](=O)[O-]
IUPAC-namn 4-methoxy-3-nitropyridin-2-amine
Molekylvikt (g/mol) 169.14
Formel vikt 169.0487