missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-(2-methylphenoxy)benzoic acid, 96%, Thermo Scientific™

Catalog Number p-7032150
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
885268-00-8
C14H13NO3
243.26
MFCD06409280
XJZGSKLLSIYBQF-UHFFFAOYSA-N
2-amino-5-2-methylphenoxy benzoic acid, benzoicacid, 2-amino-5-2-methylphenoxy
44669090
2-amino-5-(2-methylphenoxy)benzoic acid
CC1=CC=CC=C1OC1=CC(C(O)=O)=C(N)C=C1
This item is not returnable. View return policy
885268-00-8
C14H13NO3
243.26
MFCD06409280
XJZGSKLLSIYBQF-UHFFFAOYSA-N
2-amino-5-2-methylphenoxy benzoic acid, benzoicacid, 2-amino-5-2-methylphenoxy
44669090
2-amino-5-(2-methylphenoxy)benzoic acid
CC1=CC=CC=C1OC1=CC(C(O)=O)=C(N)C=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 149°C to 153°C
Quantity 1 g
Formula Weight 243.26
Percent Purity 96%
Chemical Name or Material 2-Amino-5-(2-methylphenoxy)benzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'