Läs mer
2-Amino-5-[(3,4,5-trimethoxyphenyl)methyl]-4(1H)-pyrimidinone, TRC
Produktkod. 30132557
Change view
Kvantitet:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30132557 25 mg
30137369 100 mg
30008824 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30132557 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA63083025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 92440-76-1
Molekylformel C14 H17 N3 O4
Molekylvikt (g/mol) 291.3
Synonym 2-Amino-5-(3,4,5-trimethoxybenzyl)pyrimidin-4-ol, Trimethoprim Imp. D (EP)
IUPAC-namn 2-amino-5-[(3,4,5-trimethoxyphenyl)methyl]pyrimidin-4-ol
LEDER COc1cc(Cc2cnc(N)nc2O)cc(OC)c1OC
Specifikationer
Kemiskt namn eller material 2-Amino-5-(3,4,5-trimethoxybenzyl)pyrimidin-4-ol
Analyt- eller komponentnamn 2-Amino-5-(3,4,5-trimethoxybenzyl)pyrimidin-4-ol
Anteckningar om renhetsgrad HPLC
Frakt skick Room Temperature
InChI formel InChI=1S/C14H17N3O4/c1-19-10-5-8(6-11(20-2)12(10)21-3)4-9-7-16-14(15)17-13(9)18/h5-7H,4H2,1-3H3,(H3,15,16,17,18)
Lösningstyp Neat
Industry Type Pharmaceutical
Kvantitet 25 mg
Formel vikt 291.1219
Procent renhet >95%