Läs mer
2-Amino-5-(4-hydroxy-3,5-dimethoxybenzyl)pyrimidin-4-ol, Mikromol™
Produktkod. 30051781
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30051781 100 mg
30050702 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30051781 Leverantör Mikromol Leverantörsnummer MM0093.05
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2-Amino-5-(4-hydroxy-3,5-dimethoxybenzyl)pyrimidin-4-ol
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-Amino-5-(4-hydroxy-3,5-dimethoxybenzyl)pyrimidin-4-ol
CAS 72920-13-9
Analyt- eller komponentnamn 2-Amino-5-(4-hydroxy-3,5-dimethoxybenzyl)pyrimidin-4-ol
Frakt skick Room Temperature
Molekylformel C13 H15 N3 O4
Certifikat och efterlevnad ISO 17025
Synonym 2-Amino-5-(4-hydroxy-3,5-dimethoxybenzyl)pyrimidin-4-ol
InChI formel InChI=1S/C13H15N3O4/c1-19-9-4-7(5-10(20-2)11(9)17)3-8-6-15-13(14)16-12(8)18/h4-6,17H,3H2,1-2H3,(H3,14,15,16,18)
LEDER COc1cc(Cc2cnc(N)nc2O)cc(OC)c1O
IUPAC-namn 2-amino-5-[(4-hydroxy-3,5-dimethoxyphenyl)methyl]pyrimidin-4-ol
Molekylvikt (g/mol) 277.28
Kvantitet 100 mg
Formel vikt 277.1063 g/mol