Läs mer
2-Amino-5,6-dichloro-3(4H)-quinazoline Acetic Acid (Anagrelide Impurity B), TRC
Produktkod. 30311667
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30311667 25 mg
1 options
Produktkod. 30311667 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA60474025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-Amino-5,6-dichloro-3(4H)-quinazoline Acetic Acid (Anagrelide Impurity B)
CAS 1159977-03-3
Molekylformel C10H9Cl2N3O2
Kvantitet 25 mg
Synonym USP Anagrelide Related Impurity B, Anagrelide USP Related Impurity B
InChI formel InChI=1S/C10H9Cl2N3O2/c11-6-1-2-7-5(9(6)12)3-15(4-8(16)17)10(13)14-7/h1-2H,3-4H2,(H2,13,14)(H,16,17)
LEDER NC1=Nc2ccc(Cl)c(Cl)c2CN1CC(=O)O
IUPAC-namn 2-(2-amino-5,6-dichloro-4H-quinazolin-3-yl)acetic acid
Molekylvikt (g/mol) 274.1
Formel vikt 273.0072