Läs mer
(2-Amino-5,6-dichloroquinazolin-3(4H)-yl)acetic Acid Hydrochloride, Mikromol™
Produktkod. 17991346
Change view
Kvantitet:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17991346 100 mg
17924076 25 mg
30460737 4 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17991346 Leverantör Mikromol Leverantörsnummer MM3250.04
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(2-Amino-5,6-dichloroquinazolin-3(4H)-yl)acetic Acid Hydrochloride
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (2-Amino-5,6-dichloroquinazolin-3(4H)-yl)acetic Acid Hydrochloride
CAS 2203514-95-6
Analyt- eller komponentnamn (2-Amino-5,6-dichloroquinazolin-3(4H)-yl)acetic Acid Hydrochloride
Frakt skick Room Temperature
Molekylformel C10 H9 Cl2 N3 O2 . Cl H
Certifikat och efterlevnad ISO 17025
Synonym (2-Amino-5,6-dichloroquinazolin-3(4H)-yl)acetic Acid Hydrochloride, Anagrelide Related Compound B (USP), Anagrelide RC B, Anagrelide Related Compound B as Hydrochloride
InChI formel InChI=1S/C10H9Cl2N3O2.ClH/c11-6-1-2-7-5(9(6)12)3-15(4-8(16)17)10(13)14-7;/h1-2H,3-4H2,(H2,13,14)(H,16,17);1H
LEDER Cl.NC1=Nc2ccc(Cl)c(Cl)c2CN1CC(=O)O
IUPAC-namn 2-(2-amino-5,6-dichloro-4H-quinazolin-3-yl)acetic acid;hydrochloride
Molekylvikt (g/mol) 310.56
Kvantitet 100 mg
Formel vikt 308.9839 g/mol