Läs mer
2-Amino-5-chloro-3-methylbenzoic Acid, TRC
Produktkod. 30312729
Change view
Kvantitet:
5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30312729 5 g
30310045 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30312729 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA6173735G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-Amino-5-chloro-3-methylbenzoic Acid
CAS 20776-67-4
Molekylformel C8H8NO2Cl
Kvantitet 5 g
Synonym 2-Amino-5-chloro-3-methylbenzoic Acid, 2-Amino-3-methyl-5-chlorobenzoic Acid, 3-Methyl-5-chloroanthranilic Acid
InChI formel InChI=1S/C8H8ClNO2/c1-4-2-5(9)3-6(7(4)10)8(11)12/h2-3H,10H2,1H3,(H,11,12)
LEDER Cc1cc(Cl)cc(C(=O)O)c1N
IUPAC-namn 2-amino-5-chloro-3-methylbenzoic acid
Molekylvikt (g/mol) 185.6
Formel vikt 185.0244