missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-chlorobenzaldehyde, 97%, Thermo Scientific Chemicals

Catalog Number p-7047070
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
20028-53-9
C7H6ClNO
155.581
MFCD02093936
KUIFMGITZFDQMP-UHFFFAOYSA-N
pubchem7364, acmc-209vm9, 3-chloro-6-aminobenzaldehyde, 2-amino-5-chloro-benzaldehyde, 2-amino-5-chlorobenz-aldehyde, 5-chloroanthranilaldehyde
2773898
2-amino-5-chlorobenzaldehyde
C1=CC(=C(C=C1Cl)C=O)N
This item is not returnable. View return policy
20028-53-9
C7H6ClNO
155.581
MFCD02093936
KUIFMGITZFDQMP-UHFFFAOYSA-N
pubchem7364, acmc-209vm9, 3-chloro-6-aminobenzaldehyde, 2-amino-5-chloro-benzaldehyde, 2-amino-5-chlorobenz-aldehyde, 5-chloroanthranilaldehyde
2773898
2-amino-5-chlorobenzaldehyde
C1=CC(=C(C=C1Cl)C=O)N

2-Amino-5-Chlorobenzaldehyde is used as a raw material in organic synthesis. It also serves as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-5-Chlorobenzaldehyde is used as a raw material in organic synthesis. It also serves as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 68°C to 74°C
Quantity 1 g
Beilstein 2716096
Sensitivity Air Sensitive
Solubility Information Slightly soluble in water.
Formula Weight 155.58
Percent Purity 97%
Chemical Name or Material 2-Amino-5-chlorobenzaldehyde

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'