Learn More
2-Amino-5-fluoropyridine, 95%, Thermo Scientific™
Catalog Number p-3777281
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
21717-96-4
C5H5FN2
112.11
MFCD01861120
YJTXQLYMECWULH-UHFFFAOYSA-N
2-amino-5-fluoropyridine, 5-fluoro-2-pyridinamine, 5-fluoro-2-pyridineamine, 5-fluoro-pyridin-2-ylamine, 5-fluoro-2-pyridylamine, 2-amino 5-fluoropyridine, 2-pyridinamine, 5-fluoro, 5-fluoro-2-aminopyridine, 2-amino-5-fluoro pyridine, 5-fluoropyridimine
2737701
5-fluoropyridin-2-amine
NC1=CC=C(F)C=N1
21717-96-4
C5H5FN2
112.11
MFCD01861120
YJTXQLYMECWULH-UHFFFAOYSA-N
2-amino-5-fluoropyridine, 5-fluoro-2-pyridinamine, 5-fluoro-2-pyridineamine, 5-fluoro-pyridin-2-ylamine, 5-fluoro-2-pyridylamine, 2-amino 5-fluoropyridine, 2-pyridinamine, 5-fluoro, 5-fluoro-2-aminopyridine, 2-amino-5-fluoro pyridine, 5-fluoropyridimine
2737701
5-fluoropyridin-2-amine
NC1=CC=C(F)C=N1
CAS Min % 94.0
CAS Max % 100.0
Melting Point 93°C to 97°C
Color Orange to White
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 112.11
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 2-Amino-5-fluoropyridine
Show More Show Less
compliance-icons
Product Identifier
  • 2-Amino-5-fluoropyridine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.