missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-methylbenzoic acid, 97%, Thermo Scientific Chemicals

Catalog Number p-3777287
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Plastic bottle
This item is not returnable. View return policy
2941-78-8
C8H9NO2
151.17
MFCD00007909
NBUUUJWWOARGNW-UHFFFAOYSA-N
5-methylanthranilic acid, 6-amino-m-toluic acid, 2-amino-5-methyl-benzoic acid, benzoic acid, 2-amino-5-methyl, m-toluic acid, 6-amino, 2-amino-5-methylbenzoicacid, unii-418xaj22v2, 2-amino-5-methyl benzoic acid, amino 2--5-methylbenzoic acid, pubchem4954
76255
2-amino-5-methylbenzoic acid
CC1=CC=C(N)C(=C1)C(O)=O
This item is not returnable. View return policy
2941-78-8
C8H9NO2
151.17
MFCD00007909
NBUUUJWWOARGNW-UHFFFAOYSA-N
5-methylanthranilic acid, 6-amino-m-toluic acid, 2-amino-5-methyl-benzoic acid, benzoic acid, 2-amino-5-methyl, m-toluic acid, 6-amino, 2-amino-5-methylbenzoicacid, unii-418xaj22v2, 2-amino-5-methyl benzoic acid, amino 2--5-methylbenzoic acid, pubchem4954
76255
2-amino-5-methylbenzoic acid
CC1=CC=C(N)C(=C1)C(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 174.0°C to 177.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula H2NC6H3(CH3)CO2H
Quantity 1 g
Beilstein 14, 481
Formula Weight 151.16
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 2-Amino-5-methylbenzoic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2-Amino-5-methylbenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'