Läs mer
2-Amino-5-methylphenyboronic Acid Pinacol Ester, TRC
Produktkod. 30336041
Change view
Kvantitet:
100 mg
Product Code. Kvantitet
30336041 100 mg
1 options
Product Code. 30336041 Supplier Toronto Research Chemicals Supplier No. TRCA634368100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Kemiskt namn eller material 2-Amino-5-methylphenyboronic Acid Pinacol Ester
CAS 948592-80-1
Molekylformel C13H20BNO2
Kvantitet 100 mg
InChI formel InChI=1S/C13H20BNO2/c1-9-6-7-11(15)10(8-9)14-16-12(2,3)13(4,5)17-14/h6-8H,15H2,1-5H3
LEDER Cc1ccc(N)c(c1)B2OC(C)(C)C(C)(C)O2
IUPAC-namn 4-methyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Molekylvikt (g/mol) 233.11
Formel vikt 233.1587