Learn More
2-Amino-6-[(4-fluorobenzyl)-amino]-3-nitropyridine, TRC
Product Code. 30315532
Change view
Click to view available options
Kvantitet:
500 mg
Product Code. Kvantitet
30315532 500 mg
1 options
Product Code. 30315532 Supplier Toronto Research Chemicals Supplier No. TRCA618470500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Kemiskt namn eller material 2-Amino-6-[(4-fluorobenzyl)-amino]-3-nitropyridine
CAS 33400-49-6
Molekylformel C12H11FN4O2
Kvantitet 500 mg
Synonym N6-[(4-Fluorophenyl)methyl]-3-nitro-2,6-pyridinediamine
InChI formel InChI=1S/C12H11FN4O2/c13-9-3-1-8(2-4-9)7-15-11-6-5-10(17(18)19)12(14)16-11/h1-6H,7H2,(H3,14,15,16)
LEDER Nc1nc(NCc2ccc(F)cc2)ccc1[N+](=O)[O-]
IUPAC-namn 6-N-[(4-fluorophenyl)methyl]-3-nitropyridine-2,6-diamine
Molekylvikt (g/mol) 262.24
Formel vikt 262.0866