missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-bromo-3-nitropyridine, 97%, Thermo Scientific™

Catalog Number p-4812744
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
84487-04-7
C5H4BrN3O2
218.01
YRXZIHANXVKUHP-UHFFFAOYSA-N
12097745
6-bromo-3-nitropyridin-2-amine
C1=CC(=NC(=C1[N+](=O)[O-])N)Br
This item is not returnable. View return policy
84487-04-7
C5H4BrN3O2
218.01
YRXZIHANXVKUHP-UHFFFAOYSA-N
12097745
6-bromo-3-nitropyridin-2-amine
C1=CC(=NC(=C1[N+](=O)[O-])N)Br
Melting Point 203.0°C to 204.0°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Synonym 2-amino-6-bromo-3-nitropyridine, 6-bromo-3-nitro-2-pyridinamine, 2-pyridinamine, 6-bromo-3-nitro, 6-bromo-3-nitro-pyridin-2-ylamine, 6-bromo-3-nitro-2-pyridylamine, 2-pyridinamine,6-bromo-3-nitro, acmc-209puv, chempacific 38154, 6-amino-2-bromo-5-nitropyridine, 2-amino-3-nitro-6-bromopyridine
Formula Weight 218.01
Percent Purity 97%
Chemical Name or Material 2-Amino-6-bromo-3-nitropyridine
compliance-icons
Product Identifier
  • 2-Amino-6-bromo-3-nitropyridine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'