missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-chlorobenzoic acid, 97+%, Thermo Scientific Chemicals

Catalog Number p-7081077
Click to view available options
Quantity:
5 g
10 g
25 g
This item is not returnable. View return policy
2148-56-3
C7H6ClNO2
171.58
MFCD00051530
SZCPTRGBOVXVCA-UHFFFAOYSA-N
6-chloroanthranilic acid, benzoic acid, 2-amino-6-chloro, 2-amino-6-chloro-benzoic acid, 6-chloranthranilsaure, pubchem9876, acmc-1cngq, 6-chloro anthranilic acid, 2-carboxy-3-chloroaniline, 2amino-6-chlorobenzoic acid, ksc201s6b
75071
2-amino-6-chlorobenzoic acid
C1=CC(=C(C(=C1)Cl)C(=O)O)N
This item is not returnable. View return policy
2148-56-3
C7H6ClNO2
171.58
MFCD00051530
SZCPTRGBOVXVCA-UHFFFAOYSA-N
6-chloroanthranilic acid, benzoic acid, 2-amino-6-chloro, 2-amino-6-chloro-benzoic acid, 6-chloranthranilsaure, pubchem9876, acmc-1cngq, 6-chloro anthranilic acid, 2-carboxy-3-chloroaniline, 2amino-6-chlorobenzoic acid, ksc201s6b
75071
2-amino-6-chlorobenzoic acid
C1=CC(=C(C(=C1)Cl)C(=O)O)N

2-Amino-6-chlorobenzoic acid is used as intermediate for medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-6-chlorobenzoic acid is used as intermediate for medicine.

Solubility
Very soluble in water. Soluble in methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, strong acids and bases.
TRUSTED_SUSTAINABILITY
Melting Point 149°C to 152°C
Quantity 5 g
Beilstein 2804041
Solubility Information Very soluble in water. Soluble in methanol.
Formula Weight 171.58
Percent Purity 98%
Chemical Name or Material 2-Amino-6-chlorobenzoic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'