missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-chloropurine-9-riboside, 99%, Thermo Scientific™

Catalog Number p-7077210
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
2004-07-1
C10H12ClN5O4
301.69
TXWHPSZYRUHEGT-UHFFFAOYNA-N
This item is not returnable. View return policy
2004-07-1
C10H12ClN5O4
301.69
TXWHPSZYRUHEGT-UHFFFAOYNA-N
Melting Point 165.0°C to 175.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Specific Rotation -35
Synonym benzoic acid, 4-chlorophenyl ester, p-chlorophenol benzoate, p-chlorophenyl benzoate, benzoic acid 4-chlorophenyl ester, benzoic acid, p-chlorophenyl ester, 4-chlorophenylbenzoate, acmc-1cina, cbmicro_005244, 4-chlorophenyl benzoate
Solubility Information Solubility in water: .. Other solubilities: soluble in dmso,methanol
SMILES NC1=NC(Cl)=C2N=CN(C3OC(CO)C(O)C3O)C2=N1
IUPAC Name (4-chlorophenyl) benzoate
PubChem CID 347761
Formula Weight 301.69
Percent Purity 99%
Chemical Name or Material 2-Amino-6-chloropurine-9-riboside
Show More Show Less
compliance-icons
Product Identifier
  • 2-Amino-6-chloropurine-9-riboside
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'