missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-chloropyrazine, 95%, Thermo Scientific Chemicals

Catalog Number 15498518
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
33332-28-4
C4H4ClN3
129.547
MFCD00055024
JTPXVCKCLBROOJ-UHFFFAOYSA-N
2-amino-6-chloropyrazine, 2-chloro-6-aminopyrazine, 2-pyrazinamine, 6-chloro, pyrazinamine, 6-chloro, 6-chloro-pyrazin-2-ylamine, 6-chloro-2-pyrazinamine, 6-chloropyrazin-2-ylamine, 2-amino-6-chloro-pyrazine, 6-chloropyrazine-2-ylamine, 6-amino-2-chloropyrazine
118458
6-chloropyrazin-2-amine
C1=C(N=C(C=N1)Cl)N
This item is not returnable. View return policy
33332-28-4
C4H4ClN3
129.547
MFCD00055024
JTPXVCKCLBROOJ-UHFFFAOYSA-N
2-amino-6-chloropyrazine, 2-chloro-6-aminopyrazine, 2-pyrazinamine, 6-chloro, pyrazinamine, 6-chloro, 6-chloro-pyrazin-2-ylamine, 6-chloro-2-pyrazinamine, 6-chloropyrazin-2-ylamine, 2-amino-6-chloro-pyrazine, 6-chloropyrazine-2-ylamine, 6-amino-2-chloropyrazine
118458
6-chloropyrazin-2-amine
C1=C(N=C(C=N1)Cl)N

2-Amino-6-chloropyrazine is used in the preparation of A2B adenosine receptor antagonists and other biologically active compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-6-chloropyrazine is used in the preparation of A2B adenosine receptor antagonists and other biologically active compounds.

Solubility
Soluble in dimethyl sulfoxide and methanol.

Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Quantity 5 g
Solubility Information Soluble in dimethyl sulfoxide and methanol.
Formula Weight 129.55
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2-Amino-6-chloropyrazine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'