missing translation for 'onlineSavingsMsg'
Learn More

2-amino-6-jodopurin, TRC

Product Code. 30309503
Change view
Click to view available options
Kvantitet:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30309503 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30309503 Supplier Toronto Research Chemicals Supplier No. TRCA6117215G
 Begränsad artikel
Beräknad leverans: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Högrena organiska molekyler och analytiska standarder, strategiskt levererade världen över för att möjliggöra innovation och kommersiell framgång.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material 2-Amino-6-iodopurine
CAS 19690-23-4
Molekylformel C5H4IN5
Kvantitet 5 g
Synonym 6-Iodo-9H-purin-2-amine, NSC 45154
InChI formel InChI=1S/C5H4IN5/c6-3-2-4(9-1-8-2)11-5(7)10-3/h1H,(H3,7,8,9,10,11)
LEDER IC1=C2N=CN=C2NC(=N)N1
IUPAC-namn 6-jod-1,3-dihydropurin-2-imin
Molekylvikt (g/mol) 261.02
Formel vikt 260.9511
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.