missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-methylpyrazine, 97%, Thermo Scientific™

Catalog Number 15480129
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
5521-56-2
C5H7N3
109.132
MFCD08235193
UAOOJJPSCLNTOP-UHFFFAOYSA-N
2-amino-6-methylpyrazine, pyrazinamine, 6-methyl, 6-methylpyrazine-2-ylamine, 6-methyl-2-pyrazinamine, 2-pyrazinamine,6-methyl, 2-amino-6-methyl pyrazine, 6-methyl-pyrazin-2-ylamine, acmc-1am75, 6-methyl-pyrazinamine, 6-methylpyrazin-2-ylamine
12236939
6-methylpyrazin-2-amine
CC1=CN=CC(=N1)N
This item is not returnable. View return policy
5521-56-2
C5H7N3
109.132
MFCD08235193
UAOOJJPSCLNTOP-UHFFFAOYSA-N
2-amino-6-methylpyrazine, pyrazinamine, 6-methyl, 6-methylpyrazine-2-ylamine, 6-methyl-2-pyrazinamine, 2-pyrazinamine,6-methyl, 2-amino-6-methyl pyrazine, 6-methyl-pyrazin-2-ylamine, acmc-1am75, 6-methyl-pyrazinamine, 6-methylpyrazin-2-ylamine
12236939
6-methylpyrazin-2-amine
CC1=CN=CC(=N1)N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 109.13
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-Amino-6-methylpyrazine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'