Läs mer
2-Amino-7-(chloromethyl)pteridin-4(1H)-one (>90%), TRC
Produktkod. 30240903
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30240903 50 mg
30240869 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30240903 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA60335050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1391194-56-1
Molekylformel C7 H6 Cl N5 O
Molekylvikt (g/mol) 211.61
Synonym 2-Amino-7-(chloromethyl)pteridin-4(1H)-one, 4(3H)-Pteridinone, 2-amino-7-(chloromethyl)-, 2-Amino-7-(chloromethyl)-4(3H)-pteridinone
IUPAC-namn 2-amino-7-(chloromethyl)-1H-pteridin-4-one
LEDER NC1=NC(=O)c2ncc(CCl)nc2N1
Specifikationer
Kemiskt namn eller material 2-Amino-7-(chloromethyl)pteridin-4(1H)-one
Analyt- eller komponentnamn 2-Amino-7-(chloromethyl)pteridin-4(1H)-one
Frakt skick Room Temperature
InChI formel InChI=1S/C7H6ClN5O/c8-1-3-2-10-4-5(11-3)12-7(9)13-6(4)14/h2H,1H2,(H3,9,11,12,13,14)
Lösningstyp Neat
Industry Type Forensics
Kvantitet 50 mg
Formel vikt 211.0261