Läs mer
2-Amino-N-[3-(2-chlorobenzoyl)-5-ethyl-2-thienyl]-acetamide, TRC
Produktkod. 30134905
Change view
Kvantitet:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134905 500 mg
30012243 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30134905 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA603365500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 50509-09-6
Molekylformel C15 H15 Cl N2 O2 S
Molekylvikt (g/mol) 322.81
Synonym 2-(Aminoacetamido)-3-(o-chlorobenzoyl)-5-ethylthiophene
IUPAC-namn 2-amino-N-[3-(2-chlorobenzoyl)-5-ethyl-2-thienyl]acetamide
LEDER CCc1cc(C(=O)c2ccccc2Cl)c(NC(=O)CN)s1
Specifikationer
Kemiskt namn eller material 2-Amino-N-[3-(2-chlorobenzoyl)-5-ethyl-2-thienyl]-acetamide
Kvantitet 500 mg
InChI formel InChI=1S/C15H15ClN2O2S/c1-2-9-7-11(15(21-9)18-13(19)8-17)14(20)10-5-3-4-6-12(10)16/h3-7H,2,8,17H2,1H3,(H,18,19)
Formel vikt 322.0543