Läs mer
2-Amino-N-benzylbenzamide, TRC
Produktkod. 30316522
Change view
Kvantitet:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30316522 500 mg
30319504 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30316522 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA630243500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-Amino-N-benzylbenzamide
CAS 5471-20-5
Molekylformel C14H14N2O
Kvantitet 500 mg
Synonym 2-Amino-N-(phenylmethyl)benzamide, 2-(Benzylaminocarbonyl)aniline, Anthranilic Acid Benzylamide
InChI formel InChI=1S/C14H14N2O/c15-13-9-5-4-8-12(13)14(17)16-10-11-6-2-1-3-7-11/h1-9H,10,15H2,(H,16,17)
LEDER Nc1ccccc1C(=O)NCc2ccccc2
IUPAC-namn 2-amino-N-benzylbenzamide
Molekylvikt (g/mol) 226.27
Formel vikt 226.1106